Learn More
Socorex™ Stainless Steel Bent Drenchers
Product Code. 11883193
Change view
Click to view available options
Length (Metric):
25 mm
51 mm
Unit Size:
Pack of 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11883193 25 mm Pack of 3
11843203 51 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11883193 Supplier Socorex™ Supplier No. 376.0925B
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Autoclavable Autoclavable
Product Type Bent Drencher
Material Stainless Steel
For Use With (Application) For Safe Animal Nose and Mouth Feeding
Length (Metric) 25 mm