Learn More
Bochem™ Stainless Steel Bent Tip Forceps
Description
- Sharp, bent tip
Specifications
Specifications
| Product Type | Forceps |
| Length (Metric) | 200 mm |
| Material | Stainless Steel (18/10) |
| Includes | n.a. |
| No. per Pack | 1 Pack |
| Tip Style | Bent |