Learn More
Sampling Systems™ Stainless Steel Bottle
Store liquids and powders securely with this range of high quality Stainless Steel Containers. Sampling Systems™ Stainless Steel Bottle is ideal for storing and transporting powders and liquids.
291.00 € - 299.00 €
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
15819721
|
Sampling Systems™
A836-1000 |
1000 mL | 102 mm | 178 mm |
299.00 €
Each |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15829721
|
Sampling Systems™
A836-500 |
500 mL | 83 mm | 148 mm |
291.00 €
Each |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Multi-purpose containers
- Robust construction
- Crevice free interior
- Standard lid (tri-ferrule style BS 4825-3)
- All welds ground and polished
- Complete with lid, silicone gasket (FDA Compliant) and clampband
- 316L stainless steel body and lid
Specifications
Stainless Steel (316L) | |
Tri Ferrule |