Learn More
Buerkle™ Stainless Steel Bowl, Shallow Form
Product Code. 31759365 Shop All Bürkle Products
Change view
Click to view available options
Dimensions (L x W x H):
150 x 100 x 40 mm
200 x 100 x 40 mm
230 x 120 x 50 mm
250 x 200 x 50 mm
400 x 200 x 50 mm
450 x 300 x 50 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
31759365 400 x 200 x 50 mm Each
31759358 150 x 100 x 40 mm Each
31759350 200 x 100 x 40 mm Each
31759371 230 x 120 x 50 mm Each
31759334 250 x 200 x 50 mm Each
31759346 450 x 300 x 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31759365 Supplier Buerkle™ Supplier No. 42080031
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Compact, hygienic stainless steel bowl made from high-quality 1.4301 (304) steel, offering excellent corrosion resistance and versatility
  • Rectangular, shallow stainless steel bowl with robust workmanship and high functionality
  • Made of high-quality stainless steel 1.4301(304) with excellent corrosion resistance
  • Ideal for demanding environments where hygiene is critical, such as laboratories, food processing facilities, and pharmaceutical plants
  • Low design allows easy insertion and removal of instruments or samples and improves overview and accessibility
  • Provides ample space for storing, sorting, or transporting utensils despite compact dimensions
  • Smooth, easy-to-clean surface meets high hygiene standards and supports safe working practices
  • Robust, durable, and corrosion-resistant construction
  • Low, rectangular shape optimized for ergonomic working and quick access
  • Smooth surface ensures easy cleaning and maximum hygiene
  • Versatile in use
TRUSTED_SUSTAINABILITY
Spezifikation
Capacity (Metric) 3000 mL
Dimensions (L x W x H) 400 x 200 x 50 mm
Material Stainless Steel
Type Tray
Shape Low Form, Rectangular
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.