missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, 99%, pure

Product Code. 10000770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Unit Size:
1 L
2.5 L
25 L
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10000770 2.5 L Glass Bottle 2.5 L
10579420 5 mL Glass Bottle 5mL
10090760 1 L Glass Bottle 1 L
10115633 25 L Metal Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10000770 Supplier Thermo Scientific Acros Supplier No. 115690025
  • ONLINE EXCLUSIVE PRICE
    116.00 €
    106.00 € / 2.5 L
    Save 10.00 €
    8% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127-19-5 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20.0°C
CAS Min % 98.5
CAS Max % 100.0
Density 0.9370g/mL
Quantity 2.5 L
Flash Point 70°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4370 to 1.4390
Assay Percent Range 99%
Specific Gravity 0.937
Linear Formula CH3CON(CH3)2
Vapor Pressure Vapor Pressure: 1.7mbar at 25°C
Viscosity 1.02 mPa.s (20°C)
Water 0.3% max.
Beilstein 04, 59
Fieser 01,270; 02,144; 04,165
Merck Index 15, 3248
Boiling Point 164.0°C to 166.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene
Formula Weight 87.12
Percent Purity 99%
Grade Pure
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.