Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E1, NVLAP
Product Code. 16692282 Shop All Ohaus Products
Click to view available options
Mass:
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
Unit Size:
Each
Product Code. 16692282
Brand: Ohaus™ 30390935
Only null left
Add to basket
Add to basket
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E1 Tolerances
  • OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
  • Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
  • NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NVLAP Accredited Certificate
Material Troemner Alloy 8
Mass 100 g
Tolerance 0.050 mg
Includes Protective Case