Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E2, NVLAP
Product Code. 16632382 Shop All Ohaus Products
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
Unit Size:
Each
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mass unitSize
16632382 1 mg Each
16642382 2 mg Each
16652382 5 mg Each
16662382 10 mg Each
16672382 20 mg Each
16682382 50 mg Each
16692382 100 mg Each
16602392 200 mg Each
16612392 500 mg Each
16622392 1 g Each
16632392 2 g Each
16642392 5 g Each
16652392 10 g Each
16662392 20 g Each
16672392 50 g Each
16682392 100 g Each
16692392 200 g Each
16602402 500 g Each
16612402 1 kg Each
16622402 2 kg Each
16632402 5 kg Each
16642402 10 kg Each
16652402 20 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Product Code. 16632382 Supplier Ohaus™ Supplier No. 80780284
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E2 Tolerances
  • Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
  • NVLAP Calibration certificate is accredited to ISO 17025 which is recognizable through ILAC
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished, mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NVLAP Accredited Certificate
Material 3003-H14 Aluminum
Weight (Metric) 0.020 kg
Mass 1 mg
Tolerance 0.006 mg
Includes Protective Case