Description
- Stainless steel construction
- For use with batch mills
- For cutting, grinding, or pulverizing soft, fibrous materials with a Mohs hardness up to 6
Specifications
Specifications
| Material | Stainless Steel |
| Packaging Quantity | 1 Ea. |
| Weight (Metric) | 0.007 kg |