Learn More
Schneider Gerd™ Stainless Steel Double Micro-spatula
Use this micro spatula for a variety of applications. Schneider Gerd™ Stainless Steel Double Micro-spatulas are ideal for use with a wide range of compounds.
Brand:  Schneider Gerd™ 332
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10071031
  • 7.70 € / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Spatulas are ideal for working with powders, stirring and mixing
  • Perfect for working with miniscule amounts of chemicals
  • Two different end shapes, offer increased versatility
  • Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Specifications
1 Ea.
Double Ended Micro Spatula
Stainless Steel
Double
3 mm
150 mm