missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Thermo Scientific™ Stainless Steel Dressing Scissors

Product Code. 10540042
Change view
Click to view available options
Length (Metric):
130 mm
140 mm
155 mm
Tip Style:
Round/Round
Sharp/Round
Sharp/Sharp
Unit Size:
Each
Product Code. Length (Metric) Tip Style unitSize
10540042 130 mm Round/Round Each
10088360 130 mm Sharp/Sharp Each
10208930 140 mm Sharp/Round Each
10231961 155 mm Round/Round Each
10733521 140 mm Round/Round Each
10787191 155 mm Sharp/Round Each
10777191 155 mm Sharp/Sharp Each
7 options
This item is not returnable. View return policy

Product Code. 10540042

Brand: Bochem™ 4106

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from stainless steel. Bochem™ Thermo Scientific™ Stainless Steel Dressing Scissors comes in three types.

  • Three types: round-round, sharp-round, and sharp-sharp
  • Stainless steel

Specifications

Product Type Dressing Scissors
Length (Metric) 130 mm
Material Stainless Steel
Tip Style Round/Round
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.