missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Duplex, ASTM A182 Grade F51, S31803, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Stainless Steel, Duplex, ASTM A182 Grade F51, S31803, 50g
Specifications
Specifications
| Chemical Name or Material | Cobalt/Niobium/Carbon/Tungsten/Phosphorus/Vanadium/Nitrogen/Copper/Silicon/Manganese/Nickel/Chromium/Molybdenum |
| CAS | 7439-89-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Cobalt 0.012 %/Niobium 0.012 %/Carbon 0.019 %/Tungsten 0.024 %/Phosphorus 0.0248 %/Vanadium 0.068 %/Nitrogen 0.156 %/Copper 0.24 %/Silicon 0.469 %/Manganese 1.592 %/Nickel 5.44 %/Chromium 22.57 %/Molybdenum 22.57 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction