missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Stainless Steel Fine Dosing Tap

Product Code. 17876376 Shop All asecos Products
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
17876376 Stainless Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17876376 Supplier asecos™ Supplier No. 14752
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Stainless Steel Fine Dosing Tap is suitable for safety cans and canisters 14745-14747 and 14757-14759.

  • M55 thread
  • Diameter 56 mm

Specifica

Product Type Dosing Tap
Color Gray
For Use With (Application) Safe fine dosing
For Use With (Equipment) stainless steel can
Material Stainless Steel
Quantity 1
Weight (English) 0.66 lb.
Weight (Metric) 0.3 kg
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.