missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Stainless Steel Freezer Rack
Shop All Fisherbrand Products
Click to view available options
Shelves:
Pivoting Pull-out Drawers
Vertical Locking Rods
Unit Size:
Each
Description
For cryostorage in upright freezers.
- Constructed of corrosion-resistant stainless steel.
- For Assay Plate, PCR rack and cryogenic tube box storage.
- Designed to fit in most standard upright freezers.
- Sturdy containers with 4 levels of storage, providing 16 total storage compartments.
- Wire handles on the front allow for easy removal of the rack from the freezer.
Locking Rod Freezer Rack:
- Rack includes four vertical locking rods over the side opening to secure contents in place.
- Allows stacking of short assay plates in each slot (not included):
- 96-Well Assay plates (holds 3 per compartment, up to 48 total)
- 24-Well Assay plates (holds 2 per compartment, up to 32 total)
- 96-Deep Well Assay plates (holds 1 per compartment, up to 16 total)
- 96-Well PCR racks (holds 1 per compartment, up to 16 total)
Horizontal Drawer Freezer Rack
- Horizontal drawer system allow for easy box access
- Drawers feature a stop that pivots down when fully extended
- Polycarbonate drawer handles provide a user-friendly grip
- Includes a slot and label for drawer/sample identification
- Ideal for standard boxes up to 133 x 133 x 53 mm in size
Specifications
Specifications
| Product Type | Freezer Rack |
| Array | 4 x 4 |
| Autoclavable | Not Autoclavable |
| Color | Silver |
| Holds | Boxes (2 in.) |
| Material | Stainless Steel |
| Positions | 16 |
| Shape | Rectangular |
| Shelves | Vertical Locking Rods |
| Stackable | Yes |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction