Learn More
Nickel Electro™ Clifton™ Stainless Steel Gable Lids
Product Code. 11429519 Shop All Nickel-Electro Products
Change view
Click to view available options
For Use With (Equipment):
2 and 4 L Unstirred Baths
22 and 28 L Unstirred/Shaking Baths
5 and 14 L Unstirred Baths
8 and 10 L Unstirred/Shaking Baths
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11429519 2 and 4 L Unstirred Baths Each
11449519 5 and 14 L Unstirred Baths Each
12778888 22 and 28 L Unstirred/Shaking Baths Each
11439519 8 and 10 L Unstirred/Shaking Baths Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11429519 Supplier Nickel Electro™ Supplier No. SL14
Only null left
Add to basket
Add to basket
Obtain optimum results keeping samples in a water bath free from contamination and saving energy. Designed allowing for full use of whole width of a Clifton™ water bath, Clifton™ Stainless Steel Gable Lids help reduce energy consumption and running costs.
  • Designed allowing full use of whole width of a Clifton™ water bath
  • Helps reduce evaporation
  • Asists in keeping samples in a water bath free from contamination and saves energy
  • Allows extended use of the bath, right into the edges of the tank
  • Lid design ensures all condensate returns back into the bath
  • Insulated handles for safe use
  • Reduces energy consumption and running costs for your Clifton water bath
  • Lowest height of the gable being 45mm above the bath
Specifications
For Use With (Equipment) 2 and 4 L Unstirred Baths
Material Stainless Steel