missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Stainless-Steel GC Tubing

Features a smooth, clean bore for use in HPLC systems.

Brand:  Trajan Scientific and Medical™ 062406

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10572393

  • 180.00 € / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

  • Smooth and clean bore
  • Supplied in coils
  • Supplied cleaned and passivated
Specifications

Specifications

Tubing
1/32 in.
0.80 mm
Stainless Steel
Smooth and clean bore
0.50 mm
25 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers