Learn More
Trajan Scientific and Medical™ Stainless-Steel GC Tubing
Features a smooth, clean bore for use in HPLC systems.
65.70 € - 180.00 €
Product Code | Brand | Diameter (Metric) Outer | Diameter (Metric) Inner | Overall Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Diameter (Metric) Inner | Overall Length (Metric) | Price | Quantity & Availability | |||
15258723
|
Trajan Scientific and Medical™
0624173 |
3.1 mm | 2.2 mm | 2 m |
65.70 €
2m |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
15268723
|
Trajan Scientific and Medical™
062418 |
1.6 mm | 0.8 mm | 2 m |
65.70 €
2m |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||
10572393
|
Trajan Scientific and Medical™
062406 |
0.80 mm | 0.50 mm | 25 m |
180.00 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
15248723
|
Trajan Scientific and Medical™
062416 |
1.6 mm | 0.8 mm | 10 m |
138.00 €
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
10532013
|
Trajan Scientific and Medical™
0624160 |
1.6 mm | 0.8 mm | 5 m |
109.00 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||
15218723
|
Trajan Scientific and Medical™
062401 |
0.50 mm | 0.25 mm | 25 m |
172.00 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
15238723
|
Trajan Scientific and Medical™
062415 |
3.1 mm | 2.2 mm | 5 m |
109.00 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||
Description
- Smooth and clean bore
- Supplied in coils
- Supplied cleaned and passivated
Specifications
Tubing | |
Smooth and clean bore |