Learn More
DWK Life Sciences Stainless Steel Handle, for staining tray
Product Code. 11736978 Shop All DWK Life Sciences Products
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
11736978 10/Pk. Pack of 10
1 options
Product Code. 11736978 Supplier DWK Life Sciences Supplier No. 290750002
  • ONLINE EXCLUSIVE PRICE
    35.54 €
    30.94 € / Pack of 10
    Save 4.60 €
    12% Off
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Stainless steel handle allows the staining tray to be removed without direct contact with the staining solution.
Microscopy is a method of viewing objects that lie outside the resolution of the human eye and can be examined. The objects to be observed are magnified to such an extent that they can be displayed as images. For sample preparation, the objects to be examined are cut into very thin slices. These sections are then applied to an object slide and covered with a cover glass and appropriate solutions. To view the sections under the microscope, the sections on the slide must be filled in a staining trough and stained with an appropriate staining solution.
  • Can be attached separately and not permanently to the dye rack, making it easy to insert and remove the slides
  • Allows the staining rack to be safely placed in the staining box while direct contact with the staining solution can be avoided
Specifications
Autoclavable Not Autoclavable
Material Stainless Steel
For Use With (Application) Transport of staining trays; Suspension of the dye rack
No. per Pack 10/Pk.
Quantity 10/Pk.