Learn More
Welch Ilmvac™ Stainless Steel KF Reducing Adapter
Product Code. 10591593 Shop All Welch Ilmvac Products
Change view
Click to view available options
Size:
DN 25/16
DN 40/16
DN 40/25
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10591593 DN 40/25 Each
10657852 DN 25/16 Each
10132812 DN 40/16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10591593 Supplier Welch Ilmvac™ Supplier No. 701405
  • ONLINE EXCLUSIVE PRICE
    47.50 €
    45.05 € / Pack of 1
    Save 2.45 €
    5% Off
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for use with Welch Ilmvac™ Rotary Vane Pumps.
  • Stainless Steel KF Reducing Adapter
  • Available in a variety of sizes
  • Connect cold trap for intake or exhaust Welch vacuum pumps
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Quantity 1 Each
Size DN 40/25
Type KF Reducing Adapter