missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Stainless Steel Laboratory Bottle, with DIN 168-1 Thread, GL 45

Product Code. 12696167 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12696167 1500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12696167 Supplier DWK Life Sciences Supplier No. 299016006
  • ONLINE EXCLUSIVE PRICE
    669.00 €
    538.65 € / Pack of 1
    Save 130.35 €
    19% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Stainless Steel Laboratory Bottle, with DIN 168-1 Thread, GL 45 is ideal for storage where glass is unsuitable. Made from corrosion-resistant AISI 316L stainless steel with a smooth inner finish, built-in pouring lip, and full GL 45 cap compatibility.

  • Manufactured from corrosion-resistant AISI Type 316L (1.4404) stainless steel with hygienically welded seams
  • Smooth inner surface finish corresponding to IIIc (DIN 17441) with rounded inner edges for easy cleaning
  • Polished and brushed durable exterior finish
  • GL 45 thread with built-in pouring lip for clean, controlled pouring
  • Fully compatible with all GL 45 caps; stainless steel cap with PTFE sealing disc available separately
  • Standardised GL thread and matching cap system for particularly tight sealing
  • Ideal for storage applications where glass is not suitable due to breakage risk or chemical incompatibility

Specifications

Capacity (Metric) 1500 mL
Color Silver
For Use With (Application) Storage, transport and safe handling of hazardous, dangerous and valuable materials; Storage and transport of light-sensitive materials
Material Stainless Steel (316L)
Closure Included No
Capacity (English) 51 oz.
Shape Round
Product Type Storage Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.