Learn More
Amsta Rostfritt Stainless Steel Lid - Acid Proof For 603/636-10
Product Code. 30285048
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
30285048 None Each
1 options
Product Code. 30285048 Supplier Amsta Rostfritt Supplier No. 5110220
  • ONLINE EXCLUSIVE PRICE
    43.00 €
    38.60 € / Pack of 1
    Save 4.40 €
    10% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Highest quality Stainless Steel lid - acid proof For 603/636-10 Amsta bottles and double boiler bottles. Manufactured in Sweden.
  • Highest quality in stainless steel.
  • Acid proof 316L/1.4404.
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel