Learn More
Amsta Rostfritt Stainless Steel Lid - Acid Proof For 603/636-2
Product Code. 30285066
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
30285066 None Each
1 options
Product Code. 30285066 Supplier Amsta Rostfritt Supplier No. 5110132
  • ONLINE EXCLUSIVE PRICE
    29.82 €
    25.30 € / Pack of 1
    Save 4.52 €
    15% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Highest quality Stainless Steel lid - acid proof For 603/636-2 Amsta bottles and double boiler bottles. Manufactured in Sweden.
  • Highest quality in stainless steel.
  • Acid proof 316L/1.4404.
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel