Learn More
Amsta Rostfritt Stainless Steel Lid - Acid Proof For 605/606-10, 605-12
Product Code. 30284970
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
30284970 None Each
1 options
Product Code. 30284970 Supplier Amsta Rostfritt Supplier No. 5110240
  • ONLINE EXCLUSIVE PRICE
    49.00 €
    44.30 € / Pack of 1
    Save 4.70 €
    9% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Highest quality Stainless Steel lid - acid proof For 605/606-10. 605-12 Amsta buckets. Manufactured in Sweden.
  • Highest quality in stainless steel.
  • Acid proof 316L/1.4404.
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel