Learn More
Amsta Rostfritt Stainless Steel Lid - Acid Proof For 608/609-25, 608/609-30
Product Code. 30284897
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
30284897 None Each
1 options
Product Code. 30284897 Supplier Amsta Rostfritt Supplier No. 5110300
  • ONLINE EXCLUSIVE PRICE
    80.94 €
    68.00 € / Pack of 1
    Save 12.94 €
    15% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Highest quality Stainless Steel lid - acid proof For 608/609-25. 608/609-30 Amsta transport container. Manufactured in Sweden.
  • Highest quality in stainless steel.
  • Acid proof 316L/1.4404.
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel