Learn More
Amsta Rostfritt Stainless Steel Lid For 603/636-1
Product Code. 30284949
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
30284949 None Each
1 options
Product Code. 30284949 Supplier Amsta Rostfritt Supplier No. 5010100
  • ONLINE EXCLUSIVE PRICE
    21.00 €
    18.60 € / Pack of 1
    Save 2.40 €
    11% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Highest Quality Stainless Steel lid For 603/636-1 Amsta bottles and double boiler bottles. Manufactured in Sweden.
  • Stainless steel 304 /1.4301
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel