missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Lids For Test Sieve

Product Code. 11507033 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Outer:
100 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
11507033 200 mm Each
11592573 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11507033
  • ONLINE EXCLUSIVE PRICE
    32.95 €
    29.76 € / Pack of 1
    Save 3.19 €
    9% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Providing a secure closure to Fisherbrand™ Stainless Steel Test Sieves.

  • Available in 100 and 200mm diameter sizes
  • Conforms to BS 410: Part 1: 2000, ISO 3310: Part 1: 2000

Specifications

Quantity 1 Ea.
Product Type Sieve Cover
Material Stainless Steel
For Use With (Application) Providing closure to test sieves
Diameter (Metric) Outer 200 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.