missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Custom Kel-F Hub Needles

Product Code. 10516985
Click to view available options
Needle Gauge:
10 G
14 G
16 G
17 G
20 G
22 G
22s G
25 G
28 G
30 G
Needle Point Style:
2, 3, or 4
5 or AS
Custom
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 10516985

Brand: Hamilton™ 775201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Design controlled by a series of universal standards which guarantees compatibility between manufacturers.

  • Available in various gauges
  • Provided in packs of 6
  • Custom point style
  • Autoclavable
  • Gas sterilizable
  • For Kel-F hubs
  • Custom length
  • Stainless steel Luer tip

Specifications

Autoclavable Not Autoclavable
Material Stainless Steel
Needle Gauge 10 G
Diameter (Metric) Inner 2.7 mm
Diameter (Metric) Outer 3.4 mm
Product Type Luer Lock Kel-F Hub Needle
Length (Metric) Needle 9.5 to 304 mm
Needle Point Style 5 or AS
For Use With (Equipment) LT and TLL Syringes and Connectors
Connection Type Standard Tapered
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.