Learn More
Gilson™ Stainless Steel MicroTee Connector Assembly
Product Code. 12982029
Change view
Click to view available options
Packaging Quantity:
1 Each
Unit Size:
Each
Product Code. Packaging Quantity unitSize
12982029 1 Each Each
1 options
Product Code. 12982029 Supplier Gilson™ Supplier No. 0202513
Add to basket
Add to basket
Specifications
Swept Volume 84 nL
Packaging Quantity 1 Each
Includes 3 x Pk.-152 and 3 x P-416 BLK fittings
Thread Size 5/16-24
Description MicroTee Connector Assembly
For Use With For 360 um Tubing O.D.
Material Stainless Steel
Pressure 115,000 psi