Learn More
BRAND™ Stainless Steel Piston Rod
Product Code. 10473901 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
1, 2, 5 μL
10 μL
100 μL
20 and 25 μL
200 μL
50 μL
Unit Size:
Pack of 3
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10473901 20 and 25 μL Pack of 3
11813950 200 μL Pack of 3
10727981 50 μL Pack of 3
10231931 100 μL Pack of 3
10251881 10 μL Pack of 3
10799462 1, 2, 5 μL Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10473901 Supplier BRAND™ Supplier No. 701932
Only null left
Add to basket
Add to basket
Designed for capacities greater than 20μL.
  • DE-M marking
  • Provided with seal
  • Supplied in packs of 3
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DE-M
Material Stainless Steel
Volume (Metric) 20 and 25 μL
For Use With (Equipment) Transferpettor capacity 20, 25μL
Quantity 3/Pk.