Learn More
Kap-Uts Culture Tube Closures
Product Code. 15215637 Shop All Cole-Parmer Products
Click to view available options
Size (Metric):
13 mm
16 mm
Unit Size:
Pack of 144
Product Code. 15215637
Brand: Kap-Uts 0450001
Only null left
Add to basket
Add to basket
These Culture Tube Closures ensure the viability of microbiological cultures by protecting solid and broth media from excessive evaporation. They fit snugly on rimless culture tubes and flasks while allowing for gas exchange and ventilation.
  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.
Specifications
Product Type Culture Tube Closure
Autoclavable Autoclavable
Material Stainless Steel
Quantity 144 Pk.
Size (English) 0.6 in.
Size (Metric) 16 mm
Tube Material Stainless Steel