missing translation for 'onlineSavingsMsg'
Learn More

Kap-Uts Culture Tube Closures

Product Code. 15245376 Shop All Cole-Parmer Products
Change view
Click to view available options
Size (Metric):
13 mm
16 mm
Unit Size:
Pack of 144
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Unit Size
15245376 13 mm Pack of 144
15215637 16 mm Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15245376 Supplier Kap-Uts Supplier No. 0450000
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These Culture Tube Closures ensure the viability of microbiological cultures by protecting solid and broth media from excessive evaporation. They fit snugly on rimless culture tubes and flasks while allowing for gas exchange and ventilation.

  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.

Specifications

Product Type Culture Tube Closure
Autoclavable Autoclavable
Material Stainless Steel
Quantity 144 Pk.
Size (English) 0.5 in.
Size (Metric) 13 mm
Tube Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.