Learn More
BINDER™ Reinforced Rack
Product Code. 15621846 Shop All BINDER Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 15621846
Brand: BINDER™ 80120674
Only null left
Add to basket
Add to basket
Material: Stainless Steel; For Use With: KBF 720, KBF P 720, KBF LQC 720, KB 720, KBW 720, KBWF 720, KMF 720
  • Manufactured from stainless steel
  • With one set of securing elements (4 pieces)
  • Maximum load of 70kg
Specifications
Description Rack, reinforced - stainless steel, with fasteners (1 set of 4)
For Use With (Equipment) BINDER Chambers KBF 720, KBF P 720, KBF LQC 720, KB 720, KBW 720, KBWF 720, KMF 720
Material Stainless Steel
Type Rack
Includes 1 Set of four fasteners