Learn More
Swann-Morton™ Stainless Steel Scalpel Blades, 100-pack
Product Code. 12991635
Change view
Click to view available options
Size:
# 10
# 10A
# 11
# 15
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12991635 # 15 Pack of 100
12398009 # 10 Pack of 100
12981635 # 11 Pack of 100
12971635 # 10A Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12991635 Supplier Swann-Morton™ Supplier No. 0305
  • ONLINE EXCLUSIVE PRICE
    23.70 €
    21.55 € / Pack of 100
    Save 2.15 €
    9% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Operate using Swann Morton™ Stainless Steel Scalpel Blades to ensure precision;
  • Sterile
  • Stainless Steel
Specifications
Blade Material Stainless Steel
Product Type Scalpel Blade
Size # 15
Length (Metric) 15 mm
For Use With (Equipment) The # 15 Blade fit Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L
Certifications/Compliance BS EN 7740
Quantity 10/Pk.