Description
PTFE coated forceps, providing a non-stick surface to which very few solid or liquid substances will permanently adhere.
- Chemically resistant and non-contaminating
- Non-protein binding and providing bio-inert surfaces
- No critical quantities lost due to sticking
- No build up on surfaces
- Ideal for use under severe laboratory conditions
- Excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized
- With guide pin
Specifications
Specifications
| Product Type | Forceps |
| Material | PTFE-coated Stainless Steel |
| Height (Metric) | 130 mm |
| No. per Pack | Each |
| Style | Serrated Jaws |
| Tip Style | Sharp |