missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Stainless Steel Shipping Bottle, UN certified, with DIN 168-1 Thread, GL 45

Product Code. 12616177 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12616177 1500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12616177 Supplier DWK Life Sciences Supplier No. 299016055
  • ONLINE EXCLUSIVE PRICE
    735.00 €
    698.20 € / Pack of 1
    Save 36.80 €
    5% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Stainless Steel Shipping Bottle, UN certified, with DIN 168-1 Thread, GL 45 is designed for safe transport of dangerous liquid goods. Made from corrosion-resistant AISI 316L stainless steel with UN approval for Packing Groups I, II, and III. No additional outer packaging required.

  • Certified to UN standards for the carriage of liquids classified as dangerous goods in UN Packing Groups I (high danger), II (medium danger), and III (low danger)
  • No additional outer packaging required due to UN approval
  • Suitable for liquids with a relative density of 2.0 or less
  • Manufactured from corrosion-resistant AISI Type 316L (1.4404) stainless steel with all welded seams for hygienic construction
  • Smooth inner surface finish corresponding to IIIc (DIN 17441) with rounded inner edges for ease of cleaning
  • Polished and brushed durable exterior finish
  • GL 45 thread with built-in pouring lip; supplied complete with GL 45 stainless steel screw cap and PTFE-surfaced platinum-catalysed silicone cap liner
  • International regulations are subject to change; it is the user's responsibility to comply with all applicable laws and regulations

Specifications

Capacity (Metric) 1500 mL
Color Silver
For Use With (Application) Carriage of liquids classified as dangerous goods in UN packing Groups I (high danager), II (medium danger) and III (low danger); Storage, transport and safe handling of hazardous, dangerous and valuable materials; Storage and transport of light-sensitive materials
Material Stainless Steel (316L)
Nominal Capacity 1500 mL
Closure Included Yes
Capacity (English) 51 oz.
Shape Round
Product Type Transport Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.