missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UN55plus, 15 mm
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN55plus Oven
Unit Size:
Each
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
Specifications
| For Use With (Equipment) | UN55plus Oven |
| Type | Drip Tray |
| Material | Stainless Steel |
| Height (Metric) | Rim: 15 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering