missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Stainless Steel Upright Freezer Rack for Cryogenic Boxes

Product Code. 15250654 Shop All TENAK Products
Change view
Click to view available options
Holds:
Boxes (10)
Boxes (12)
Boxes (15)
Boxes (16)
Boxes (20)
Boxes (25)
Boxes (28)
Array:
2 x 5
3 x 5
4 x 3
4 x 4
4 x 5
5 x 3
5 x 4
5 x 5
7 x 4
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Array unitSize
15250654 Boxes (12) 4 x 3 Each
11501825 Boxes (16) 4 x 4 Each
15210674 Boxes (25) 5 x 5 Each
15290664 Boxes (20) 4 x 5 Each
15250684 Boxes (12) 3 x 5 Each
15250664 Boxes (28) 7 x 4 Each
15250704 Boxes (10) 2 x 5 Each
15210664 Boxes (20) 5 x 4 Each
15268373 Boxes (15) 5 x 3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15250654 Supplier Tenak™ Supplier No. TE23122
  • ONLINE EXCLUSIVE PRICE
    106.00 €
    104.11 € / Pack of 1
    Save 1.89 €
    1% Off
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use Tenak™ Stainless Steel Upright Freezer Racks for Cryogenic Boxes for your cryogenic storage needs. A variety of stainless steel racks accommodate box heights and quantities for laboratory freezers.

  • Suitable for all brands of upright freezers
  • Stainless steel
  • For cryogenic boxes of various capacities

Specifications

Cell Dividers Yes
Holds Boxes (12)
Material Stainless Steel
Positions 12
For Use With (Application) Fit into any freezer brand and model
For Use With (Equipment) Upright Freezers
No. per Pack 1
Quantity 1 Ea.
Depth (Metric) 419 mm
Product Type Standard Rack
Shelves 12
Height (Metric) 221 mm
Array 4 x 3
Width (Metric) 139 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.