Learn More
Swann-Morton™ Stainless Steel Surgical Scalpels
Product Code. 11768353
Change view
Click to view available options
Size:
# 10A
# 11
# 12
# 15
# 20
# 21
# 22
# 22A
# 23
# 24
Unit Size:
Pack of 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11768353 # 23 Pack of 10
12397999 # 10A Pack of 10
12308009 # 11 Pack of 10
11718353 # 12 Pack of 10
11728353 # 15 Pack of 10
11738353 # 20 Pack of 10
11758353 # 22 Pack of 10
11748353 # 21 Pack of 10
12368009 # 22A Pack of 10
11778353 # 24 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11768353 Supplier Swann-Morton™ Supplier No. 0510
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Use for convenience in user environments: on the ward, in emergency or trauma units or even out in the field with paramedics. Swann Morton™ Stainless Steel Surgical Scalpels come in single peel packs.
  • Stainless steel blade
  • Polystyrene handle
  • Individually wrapped
  • Sterile
  • Disposable
Specifications
Blade Material Stainless Steel
For Use With (Application) Used for Making Long Incisions such as an Upper Midline Incision of the Abdomen during the repair of a Perforated Gastric Ulcer
Size # 23
Disposable Single-use
Sterility Sterile
Handle Material Polystyrene