missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Stainless Steel Universal Scissors

Product Code. 10525611
Change view
Click to view available options
Length (Metric):
180 mm
200 mm
210 mm
Tip Style:
Sharp/Round
Sharp/Sharp
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tip Style Unit Size
10525611 180 mm Sharp/Round Each
10517892 200 mm Sharp/Sharp Each
10060761 210 mm Sharp/Sharp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10525611 Supplier Bochem™ Supplier No. 4151
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Temperature-resistant universal scissors with a plastic handle. Bochem™ Stainless Steel Universal Scissors are constructed of resistant and durable stainless steel.

  • Stainless steel construction.
  • Plastic handle.
  • Chemically and thermally resistant (up to 145°C).

Specifications

Product Type Universal Scissors
Length (Metric) 180 mm
Material Stainless Steel, Plastic
Tip Style Sharp/Round
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.