missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Stand

Product Code. 16373677 Shop All Brand Products
Change view
Click to view available options
Color:
Black/White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
16373677 Black/White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16373677 Supplier BRANDTECH™ BRAND™ Supplier No. 705230
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

BRAND™ Stand is an accessory part for use with HandyStep™ Touch and HandyStep™ Touch S.

  • Accessory
  • For use with HandyStep™ Touch and Touch S Electronic Repeating Pipette
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Black/White
Holds Electronic Pipette
Material Plastic
Positions 1
Shape Horizontal
Stackable No
Sterility Non-sterile
For Use With (Equipment) HandyStep Touch / Touch S
Quantity 1
Product Type Stand
Shelves None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.