Learn More
Koettermann™ Standard Cabinet for Acids and Bases, PP, 900x720x520
Product Code. 14805361
Change view
Click to view available options
Dimensions (D x W x H):
515 x 900 x 720 mm
Unit Size:
Each
Product Code. Dimensions (D x W x H) Unit Size
14805361 515 x 900 x 720 mm Each
1 options
Product Code. 14805361 Supplier Koettermann™ Supplier No. 37700011
 Restricted Item
Estimated Shipment: 27-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Koettermann™ Standard Cabinet for Acids and Bases, PP, 900x720x520 provides suitable storage to accommodate a variety of laboratory needs, but is especially suited for storing acids and bases.
  • Acid/base cabinet
  • Two doors
  • Three drawers
  • Fan
Specifications
Dimensions (D x W x H) 515 x 900 x 720 mm
Includes 3 Drawers, Fan
No. of Doors 2
Product Type Acids and Bases Cabinet