Learn More
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco™ Injectors
Product Code. 10631714 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10631714 500 μL Each
10324133 5 μL Each
10110743 10 μL Each
10715564 25 μL Each
10171073 50 μL Each
10160503 100 μL Each
10120553 250 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10631714 Supplier Thermo Scientific™ Supplier No. 365JL281
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
TRUSTED_SUSTAINABILITY
Specifications
Description 500uL
Product Type HPLC Autosampler Syringe
Disposable Reusable
Volume (Metric) 500 μL
For Use With (Equipment) Injector
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.