missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Standard Grade Rochester Pean Forceps, Straight

Product Code. 15700439 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
18.415 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15700439 18.415 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15700439 Supplier Cole-Parmer™ Supplier No. 1081813
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Standard-grade Rochester Pean forceps are manufactured from stainless steel; hardened, tempered, and polished for a high-priced finish without the added cost. These instruments are well-suited for less critical applications.

  • Standard grade
  • Stainless steel
  • Straight end type
  • Not intended for medical applications or patient use
  • Autoclavable

Specifications

Length (English) 7.25 in.
Material Stainless Steel
Length (Metric) 18.415 cm
Product Type Forceps
Grade Standard
For Use With (Application) Well suited for less critical applications
Description Standard grade Rochester Pean straight end forceps; Autoclavable
Style Straight end
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.