missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Standard Rack for Upright Freezer, 50 mm Box Height, with Boxes

Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE23130B

Code : NEW

Additional Details : Weight : 3.20000kg

 View more versions of this product

Product Code. 15290654

  • 137.00 € / Each
Estimated Shipment: 17-09-2024
to see stock.
Add to basket

Description

Description

  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications

Specifications

White cardboard boxes, 9 x 9 cell dividers
Boxes (12)
Upright Freezers
50 mm (Box)
3 x 4
Stainless Steel
560 x 139 x 168 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers