Learn More
Spectrum™ Standard RC Dialysis Tubing
Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes
Brand: Spectrum™ 132700
Code : LB
Additional Details : Weight : 0.14400kg
Description
Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes
- Membrane grade
- Membrane type: RC
- Molecular weight cut-off: 12 to 14K
Specifications
Dialysis Membrane | |
Regenerated Cellulose | |
0.6 in. | |
30 m | |
Each |
12 to 14 kDa | |
16 mm | |
98.4 ft. | |
2 mL | |
Some tubing rolls do not contain glycerin and do not require soaking prior to use |