missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Standard RC Dialysis Tubing

Product Code. 11465859
Change view
Click to view available options
Diameter (Metric):
6.4 mm
16 mm
20.4 mm
29 mm
48 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11465859 48 mm Each
11445859 6.4 mm Each
11455859 16 mm Each
11581060 20.4 mm Each
11591060 29 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11465859 Supplier Spectrum™ Supplier No. 132709
  • ONLINE EXCLUSIVE PRICE
    651.00 €
    353.00 € / Pack of 1
    Save 298.00 €
    45% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes

  • Membrane grade
  • Membrane type: RC
  • Molecular weight cut-off: 12 to 14K

Specifications

Product Type Dialysis Membrane
MWCO 12 to 14 kDa
Material Regenerated Cellulose
Format Tubing
Diameter (Metric) 48 mm
Diameter (English) 1.88 in.
Length (English) 50 ft.
Length (Metric) 15 m
Capacity (Metric) 18 mL
Quantity 50 Pack
For Use With (Application) Some tubing rolls do not contain glycerin and do not require soaking prior to use
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.