missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Standard RC Dialysis Tubing

Product Code. 11591060
Change view
Click to view available options
Diameter (Metric):
6.4 mm
16 mm
20.4 mm
29 mm
48 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11591060 29 mm Each
11465859 48 mm Each
11445859 6.4 mm Each
11455859 16 mm Each
11581060 20.4 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11591060 Supplier Spectrum™ Supplier No. 132706
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes

  • Membrane grade
  • Membrane type: RC
  • Molecular weight cut-off: 12 to 14K

Specifications

Product Type Spectra/Por 4 Dialysis Tubing
MWCO 12 to 14 kDa
Material Regenerated Cellulose
Diameter (Metric) 29 mm
Diameter (English) 1.14 in.
Length (English) 100 ft.
Length (Metric) 30 m
Formulation Contain glycerin as a humectant
Temperature Range (English) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantity 100 Pack
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.