Learn More
Electrothermal™ Standard Reaction Vessels
Product Code. 12514236
Change view
Click to view available options
For Use With (Equipment):
RS600 and OS6000 Series
Unit Size:
Pack of 6
Product Code. For Use With (Equipment) Unit Size
12514236 RS600 and OS6000 Series Pack of 6
1 options
Product Code. 12514236 Supplier Electrothermal™ Supplier No. 3663052
  • ONLINE EXCLUSIVE PRICE
    1540.00 €
    1201.13 € / Pack of 6
    Save 338.87 €
    22% Off
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Used with Electrothermal™ RS600 Reaction Station and Omni OS6000 Series Reaction Stations
  • For use with Electrothermal™ RS600 Reaction Station and Omni OS6000 Series Reaction Stations.
  • 250mL
  • Round Bottomed Flasks
  • 56.5mm diameter
  • Quantity: 6/PK
Specifications
For Use With (Equipment) RS600 and OS6000 Series