missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 2'-OMe-Bz-A Phosphoramidite, standard grade, serum vial bottle
Shop All Thermo Scientific ProductsDescription
Ensure quality and reliability for RNA oligonucleotide synthesis with Thermo Scientific™ Standard RNA 2'-TBDMS, 2'-OMe and 2'-F Phosphoramidites.
- Rigorously controlled manufacturing processes minimize levels of reactive P(III) impurities which could be incorporated into the oligonucleotide product and/or result in the accumulation of by-products which can be difficult to remove during purification
- NMR analysis is conducted on each lot to assure that the levels of critical impurities are within specifications
Process and Supply Chain Control:
- Processes are strictly controlled and monitored from material procurement through final QC and product release under ISO9001 standards, providing batch-to-batch reproducibility
- All standard RNA phosphoramidites are supplied with lot-specific quality control data including HPLC and phosphorous NMR analyses
- TBDMS RNA amidite chemistry is the widely accepted standard for high quality, large-scale synthesis of RNA oligonucleotides
- Specifications include limits for critical impurities
2'-OMe:
- RNA oligonucleotides synthesized using 2'-OMe modified amidites are nuclease resistant, making them well suited for in vivo, antisense and aptamer applications
2'-F:
- RNA oligonucleotides synthesized using 2'-Fluoro U and C phosphoramidites have increased thermal stability of RNA duplexes and nuclease resistance of single-stranded RNA
- These phosphoramidites can be incorporated into custom siRNA oligonucleotides for use in gene-targeting and suppression studies
Flexible Packaging:
- Common pack sizes fit most commercially available synthesizers and are in stock for immediate delivery
- Custom and bulk packaging are available
Specifications
Specifications
| Format | Powder |
| For Use With (Equipment) | AKTA Synthesizers, Applied Biosystems Synthesizers |
| Product Type | 2'-OMe-Bz-A CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Adenosine (A) |
| Reagent Features | 2'-OMe modified |
| Shipping Condition | Room Temperature |
| Purity | 98% |
| Quantity | 1 g |
| Reagent Type | Phosphoramidite |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?