missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Standard solutions for the control of photometric accuracy
- Verifies the instrument's absorbance accuracy
- For use with NanoPhotometer™ NP120/NP80/N60/N50
- CAS-No: 877-24-7
Specifications
Specifications
| Type | Non-toxic Potassium Hydrogen Phthalate Solution (PHP) |
| For Use With (Equipment) | NanoPhotometer N60 and NP80 |
| Packaging | 2 x 0.5 mL |
| Quantity | 2 Vials |
| Storage Requirements | Store at room temperature |
| Wavelength | 280 nm |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?