Learn More
Heinz Herenz Standard Tip LMS-Swab Amies
Product Code. 30178743
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 150
Product Code. Sterility Unit Size
30178743 Sterile Pack of 150
1 options
Product Code. 30178743 Supplier Heinz Herenz Supplier No. 1020203
  • ONLINE EXCLUSIVE PRICE
    139.67 €
    90.03 € / Pack of 150
    Save 49.64 €
    35% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Test tube with pink screw cap containing 1 mL of liquid Amies medium for sampling and preservation of aerobic, anaerobic and other fastidious bacteria within 48 hours.
Our transport and collection swabs contain special applicators and test tubes with different liquid media, customized for the transportation and preservation of microbiological samples.
  • The liquid medium allows the division of the sample material and thus several tests from one sample.
  • The ready to use devices are supplied in a peelpack which is easy to open.
  • The breakpoint in the shaft of the applicator enables a safe and hygienic workflow.
  • The standard size of the test tubes with screwcap simplified the use with automatic analyzers.
  • There are labels on the test tubes for a accurate sample identification.
  • The items are produced according to the conditions of GMP (Good Manufacturing Practice) and certified according to DIN EN ISO 13485. This ensures a high quality.
  • Our swabs are sterile (SAL 106), pyrogen free and noncytotoxic.
Specifications
Product Type Liquid Amies Transport System
Format Swab
Media Type Liquid Amies
Sterility Sterile
Color White
Tip Type Standard Tip